Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

282138

Sigma-Aldrich

Bis(trimethylsilyl)methane

97%

Synonym(s):

Methylenebis(trimethylsilane)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3Si]2CH2
CAS Number:
Molecular Weight:
160.40
Beilstein:
1734530
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.417 (lit.)

bp

133-134 °C (lit.)

mp

−71 °C (lit.)

density

0.751 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(C)C[Si](C)(C)C

InChI

1S/C7H20Si2/c1-8(2,3)7-9(4,5)6/h7H2,1-6H3

InChI key

GYIODRUWWNNGPI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lin Yang et al.
Journal of hazardous materials, 405, 124194-124194 (2020-10-25)
A novel CO2-responsive cellulose nanofibril aerogel as a "green" adsorbent derived from poly(methacrylic acid-co-2-(dimethylamino) ethyl methacrylate) and carboxylated cellulose nanofibrils was successfully prepared via stepwise cation-induced gelation and freeze drying method. This aerogel exhibited CO2-triggered adsorption behavior towards anionic dyes
Ariagna L Rivera-Briso et al.
International journal of biological macromolecules, 143, 1000-1008 (2019-11-18)
Poly(3-hydroxybutyrate-co-3-hydroxyvalerate) (PHBV) is a microbial biodegradable polymer with a wide range of potential industrial applications. However, its biomedical uses could increase exponentially if certain physical and biological properties were enhanced without compromising on the non-cytotoxic property of this biocompatible polymer.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service