Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

280046

Sigma-Aldrich

1-Iodoundecane

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)10I
CAS Number:
Molecular Weight:
282.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.4849 (lit.)

bp

125-130 °C/5 mmHg (lit.)

density

1.22 g/mL at 25 °C (lit.)

functional group

alkyl halide
iodo

SMILES string

CCCCCCCCCCCI

InChI

1S/C11H23I/c1-2-3-4-5-6-7-8-9-10-11-12/h2-11H2,1H3

InChI key

FKUQOQPBCHJHAP-UHFFFAOYSA-N

General description

1-Iodoundecane is one of the volatile constituent of urine of a normal male mice. The reaction of 1-iodoundecane with fluoride sources has been investigated.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reinhard Schwesinger et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 12(2), 438-445 (2005-10-01)
Several peralkylated polyaminophosphazenium cations were evaluated for the generation of novel anhydrous F- salts. Two of them have been characterized by X-ray analysis and are particularly soluble, even in apolar aprotic solvents like benzene or THF, one of them even
S Achiraman et al.
Animal reproduction science, 87(1-2), 151-161 (2005-05-12)
A previous investigation revealed that urine from normal male mice contained five unique volatile constituents; namely: 3-cyclohexene-1-methanol (I); 3-amino triazole (II); 4-ethyl phenol (III); 3-ethyl-2,7-dimethyl octane (IV); 1-iodoundecane (V). The present study was designed to find out whether the production

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service