Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

279749

Sigma-Aldrich

2-Methylaminomethyl-1,3-dioxolane

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H11NO2
CAS Number:
Molecular Weight:
117.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

98%

refractive index

n20/D 1.4404 (lit.)

density

1.029 g/mL at 25 °C (lit.)

SMILES string

CNCC1OCCO1

InChI

1S/C5H11NO2/c1-6-4-5-7-2-3-8-5/h5-6H,2-4H2,1H3

InChI key

ZCCPVTVGEQLONB-UHFFFAOYSA-N

Application

2-Methylaminomethyl-1,3-dioxolane was used in the synthesis of N-[methyl(2-hydroxy-3,5-dimethylphenyl)]-N-methyl-N-methyl-1,3-dioxolaneamine (mpoa-H)[1].

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

136.4 °F - closed cup

Flash Point(C)

58 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jakub Wojtaszak et al.
Dalton transactions (Cambridge, England : 2003), 43(6), 2424-2436 (2013-12-07)
The reaction of MgBu2, ZnEt2 or Ca(O(i)Pr)2 with 2 eq. of three-coordinating N-[methyl(2-hydroxy-3,5-dimethylphenyl)]-N-methyl-N-methyl-1,3-oxolaneamine (mpoa-H) or N-[methyl(2-hydroxy-3,5-di-tert-butylphenyl)]-N-methyl-N-methyl-1,3-oxolaneamine (tbpoa-H) gave neutral, monomeric [Mg(mpoa)2], [Zn(mpoa)2], [Zn(tbpoa)2], and [Ca(tbpoa)2] as white powders in 58-90% yields. The resulting aminophenolates were characterized in solution by NMR

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service