Skip to Content
MilliporeSigma
All Photos(1)

Documents

279552

Sigma-Aldrich

4-Chloro-3-ethylphenol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(C2H5)OH
CAS Number:
Molecular Weight:
156.61
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

45-50 °C (lit.)

SMILES string

CCc1cc(O)ccc1Cl

InChI

1S/C8H9ClO/c1-2-6-5-7(10)3-4-8(6)9/h3-5,10H,2H2,1H3

InChI key

DVKVZPIRWWREJC-UHFFFAOYSA-N

Application

4-Chloro-3-ethylphenol is a ryanodine receptor antagonist and is commonly used as research tool and diagnostic reagent for malignant hyperthermia.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L J Janssen et al.
The American journal of physiology, 276(5 Pt 1), L744-L753 (1999-05-18)
We examined cytosolic concentration of Ca2+ ([Ca2+]i) in canine airway smooth muscle using fura 2 fluorimetry (global changes in [Ca2+]i), membrane currents (subsarcolemmal [Ca2+]i), and contractions (deep cytosolic [Ca2+]i). Acetylcholine (10(-4) M) elicited fluorimetric, electrophysiological, and mechanical responses. Caffeine (5
Aniko Varadi et al.
Diabetes, 51 Suppl 1, S190-S201 (2002-01-30)
The endoplasmic reticulum (ER) plays a pivotal role in the regulation of cytosolic Ca(2+) concentrations ([Ca(2+)](cyt)) and hence in insulin secretion from pancreatic beta-cells. However, the molecular mechanisms involved in both the uptake and release of Ca(2+) from the ER
Francisco H Andrade et al.
Investigative ophthalmology & visual science, 46(12), 4541-4547 (2005-11-24)
The ultrafast extraocular muscles necessitate tight regulation of free cytosolic Ca2+ concentration ([Ca2+]i). Mitochondrial Ca2+ influx may be fast enough for this role. In the present study, three hypotheses were tested: (1) Mitochondrial Ca2+ uptake regulates [Ca2+]i and production of
Bo Zeng et al.
British journal of pharmacology, 171(5), 1250-1259 (2014-03-29)
Depletion of the Ca(2+) store by ryanodine receptor (RyR) agonists induces store-operated Ca(2+) entry (SOCE). 4-Chloro-3-ethylphenol (4-CEP) and 4-chloro-m-cresol (4-CmC) are RyR agonists commonly used as research tools and diagnostic reagents for malignant hyperthermia. Here, we investigated the effects of
M U Gerbershagen et al.
European journal of anaesthesiology, 19(2), 135-140 (2002-05-10)
The in vitro contracture test with halothane and caffeine is the current gold standard for diagnosis of malignant hyperthermia. This test has a sensitivity of 99.0% but a specificity of only 93.6%. Therefore, an alternative drug is desirable which distinguishes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service