Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

278769

Sigma-Aldrich

3-Chlorobicyclo[3.2.1]oct-2-ene, mixture of endo and exo

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11Cl
CAS Number:
Molecular Weight:
142.63
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

form

liquid

refractive index

n20/D 1.5055 (lit.)

bp

56 °C/10 mmHg (lit.)

density

1.082 g/mL at 25 °C (lit.)

functional group

chloro

SMILES string

ClC1=C[C@H]2CC[C@H](C2)C1

InChI

1S/C8H11Cl/c9-8-4-6-1-2-7(3-6)5-8/h4,6-7H,1-3,5H2/t6-,7+/m0/s1

InChI key

XLVAWKKCOIWHBT-NKWVEPMBSA-N

Application

3-Chlorobicyclo[3.2.1]oct-2-ene has been used in the preparation of bicyclo[3.2.1]oct-2-ene by Birch reduction.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

138.2 °F - closed cup

Flash Point(C)

59 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nuclear magnetic resonance studies. XXX. Hydrogen-deuterium exchange in bicyclo [3.2. 1] and-[2.2. 2] octenes through allylic and vinylic anions.
Cheng AK and Stothers JB.
Canadian Journal of Chemistry, 55(1), 50-55 (1977)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service