Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

278009

Sigma-Aldrich

Di-tert-butyltin dichloride

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3C]2SnCl2
CAS Number:
Molecular Weight:
303.84
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

bp

66 °C/3 mmHg (lit.)

mp

40-42 °C (lit.)

SMILES string

CC(C)(C)[Sn](Cl)(Cl)C(C)(C)C

InChI

1S/2C4H9.2ClH.Sn/c2*1-4(2)3;;;/h2*1-3H3;2*1H;/q;;;;+2/p-2

InChI key

PEGCFRJASNUIPX-UHFFFAOYSA-L

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organometallics, 8, 2871-2871 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service