Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

275093

Sigma-Aldrich

N,N′-Bis(2-hydroxy-α-phenylbenzylidene)ethylenediamine

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[HOC6H4C(C6H5)=NCH2-]2
CAS Number:
Molecular Weight:
420.50
MDL number:
UNSPSC Code:
12171500

Assay

99%

mp

162-163 °C (lit.)

SMILES string

Oc1ccccc1\C(=N\CC\N=C(\c2ccccc2)c3ccccc3O)c4ccccc4

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service