Skip to Content
MilliporeSigma
All Photos(2)

Documents

274593

Sigma-Aldrich

Diethylenetriaminepentaacetic acid iron(III) disodium salt hydrate

98%

Synonym(s):

Disodium iron(III) pentetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H18FeN3Na2O10 · xH2O
Molecular Weight:
490.13 (anhydrous basis)
EC Number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

SMILES string

O.[Na+].[Na+].[O-]C(=O)CN(CCN1CCN2CC(=O)O[Fe](OC(=O)C1)OC(=O)C2)CC([O-])=O

InChI

1S/C14H23N3O10.Fe.2Na.H2O/c18-10(19)5-15(1-3-16(6-11(20)21)7-12(22)23)2-4-17(8-13(24)25)9-14(26)27;;;;/h1-9H2,(H,18,19)(H,20,21)(H,22,23)(H,24,25)(H,26,27);;;;1H2/q;+3;2*+1;/p-5

InChI key

QPJPBTBEMHBPHZ-UHFFFAOYSA-I

Related Categories

Application

Diethylenetriaminepentaacetic acid iron(III) disodium salt hydrate may be used in chemical synthesis studies.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service