Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

274542

Sigma-Aldrich

2-Bromo-2′-nitroacetophenone

99%

Synonym(s):

2′-Nitrophenacyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4COCH2Br
CAS Number:
Molecular Weight:
244.04
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

55-57 °C (lit.)

functional group

bromo
ketone
nitro

SMILES string

[O-][N+](=O)c1ccccc1C(=O)CBr

InChI

1S/C8H6BrNO3/c9-5-8(11)6-3-1-2-4-7(6)10(12)13/h1-4H,5H2

InChI key

SGXUUCSRVVSMGK-UHFFFAOYSA-N

Application

2-Bromo-2′-nitroacetophenone, an electroactive derivative-forming reagent, was used as a precolumn reagent for preparing prostaglandin derivatives[1].

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G M Beck et al.
Talanta, 36(3), 373-377 (1989-03-01)
Conventional reversed-phase HPLC conditions have been optimized for resolution of a mixture containing prostaglandins PGE(1), PGE(2), PGF(1alpha), and PGF(2alpha). Electroactive derivative-forming reagents, such as p-nitrobenzyloxyamine, 2-bromo-2'-nitroacetophenone, and 2,4-dinitrophenylhydrazine have been evaluated for use as precolumn reagents for forming prostaglandin derivatives.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service