Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

274461

Sigma-Aldrich

9-Methylacridine

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H11N
CAS Number:
Molecular Weight:
193.24
EC Number:
MDL number:
UNSPSC Code:
12171500

Assay

95%

mp

117-119 °C (lit.)

SMILES string

Cc1c2ccccc2nc3ccccc13

InChI

1S/C14H11N/c1-10-11-6-2-4-8-13(11)15-14-9-5-3-7-12(10)14/h2-9H,1H3

InChI key

FLDRLXJNISEWNZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service