Skip to Content
MilliporeSigma
All Photos(1)

Documents

273880

Sigma-Aldrich

Methyl indole-4-carboxylate

99%

Synonym(s):

4-Methoxycarbonylindole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9NO2
CAS Number:
Molecular Weight:
175.18
Beilstein:
141826
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

68-71 °C (lit.)

SMILES string

COC(=O)c1cccc2[nH]ccc12

InChI

1S/C10H9NO2/c1-13-10(12)8-3-2-4-9-7(8)5-6-11-9/h2-6,11H,1H3

InChI key

WEAXQUBYRSEBJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The IR and UV spectra of methyl indole-4-carboxylate was studied.

Application

  • Reactant for preparation of tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential anticancer immunomodulators
  • Reactant for preparation of cytotoxic agents against multidrug-resistant cancer cells
  • Reactant for preparation of inhibitor for β-tryptase
  • Reactant for preparation of histamine H3 antagonists
  • Reactant for preparation of JNK3 MAP kinase inhibitors
  • Reactant for preparation of nucleosides

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isocarbostyrils. II. The Conversion of 2-Methyl-4-acyl-5-nitroisocarbostyrils to 2-Substituted Indole-4-carboxylic Acids.
Canadian Journal of Chemistry, 49(17), 2797-2802 (1971)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service