Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

272795

Sigma-Aldrich

N-Benzylideneaniline

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5N=CHC6H5
CAS Number:
Molecular Weight:
181.23
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47
To inquire about this product 272795, please contact your local MilliporeSigma office or dealer. Contact Technical Service

Assay

99%

form

powder

mp

52-54 °C (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

c1ccc(cc1)\C=N\c2ccccc2

InChI

1S/C13H11N/c1-3-7-12(8-4-1)11-14-13-9-5-2-6-10-13/h1-11H/b14-11+

InChI key

UVEWQKMPXAHFST-SDNWHVSQSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Kai Jiang et al.
Bioscience, biotechnology, and biochemistry, 82(8), 1265-1300 (2018-04-22)
Plant hormones are small molecules that play versatile roles in regulating plant growth, development, and responses to the environment. Classic methodologies, including genetics, analytic chemistry, biochemistry, and molecular biology, have contributed to the progress in plant hormone studies. In addition

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service