Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

272159

Sigma-Aldrich

4-Acetoxy-2-azetidinone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H7NO3
CAS Number:
Molecular Weight:
129.11
Beilstein:
1524738
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

38-40 °C (lit.)

solubility

formic acid: soluble 50 mg/mL, clear, yellow-orange

storage temp.

2-8°C

SMILES string

CC(=O)OC1CC(=O)N1

InChI

1S/C5H7NO3/c1-3(7)9-5-2-4(8)6-5/h5H,2H2,1H3,(H,6,8)

InChI key

OEYMQQDJCUHKQS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Acetoxy-2-azetidinone was used in the synthesis of derivatized cyclopentenes in high regio- and diastereoselectivity[1]. It was also used as a heterocyclic synthon[2] for antibiotic[3] and anti-inflammatory[4] agents.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aldrichimica Acta, 18, 95-95 (1985)
Journal of the Chemical Society. Perkin Transactions 1, 1235-1235 (1991)
Tetrahedron, 46, 2255-2255 (1990)
Cara Cesario et al.
Organic letters, 11(6), 1293-1295 (2009-02-17)
Acylnitroso-derived hetero-Diels-Alder cycloadducts are susceptible to C-O bond cleavage with Pd(0) and InI to form allylic indium(III) species. The in situ prepared allylindium compounds readily react at room temperature with Eschenmoser's salt. Allylation of 4-acetoxy-2-azetidinone provides derivatized cyclopentenes in high

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service