Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

270628

Supelco

Cyclohexane

HPLC grade, ≥99.9%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12
CAS Number:
Molecular Weight:
84.16
Beilstein:
1900225
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:

grade

HPLC grade

Quality Level

vapor density

2.9 (vs air)

vapor pressure

168.8 mmHg ( 37.7 °C)
77 mmHg ( 20 °C)

Assay

≥99.9%

autoignition temp.

500 °F

purified by

glass distillation

expl. lim.

9 %

impurities

<0.010% water

evapn. residue

<0.0005%

refractive index

n20/D 1.426 (lit.)

bp

80.7 °C (lit.)
80.7 °C

mp

4-7 °C (lit.)

density

0.779 g/mL at 25 °C (lit.)

λ

H2O reference

UV absorption

λ: 210 nm Amax: 1.00
λ: 220 nm Amax: 0.30
λ: 235 nm Amax: 0.09
λ: 250 nm Amax: 0.02
λ: 260-400 nm Amax: 0.01

SMILES string

C1CCCCC1

InChI

1S/C6H12/c1-2-4-6-5-3-1/h1-6H2

InChI key

XDTMQSROBMDMFD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-4.0 °F - closed cup

Flash Point(C)

-20 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cyclohexane oxidation continues to be a challenge.
Schuchardt ULF, et al.
Applied Catalysis A: General, 211(1), 1382-1386 (2012)
Omid D Rahmanian et al.
Microfluidics and nanofluidics, 18(5-6), 1045-1053 (2015-05-15)
A simple and reliable method for fabricating single-use normally closed burst valves in thermoplastic microfluidic devices is presented, using a process flow that is readily integrated into established workflows for the fabrication of thermoplastic microfluidics. An experimental study of valve
Eagleson M.
Concise Encyclopedia Chemistry, 168-168 (1994)
Lei Zhou et al.
Nature communications, 6, 6938-6938 (2015-04-25)
The identification of potential diagnostic markers and target molecules among the plethora of tumour oncoproteins for cancer diagnosis requires facile technology that is capable of quantitatively analysing multiple biomarkers in tumour cells and tissues. Diagnostic and prognostic classifications of human
Amanda L Pitts et al.
Journal of the American Chemical Society, 136(24), 8614-8625 (2014-05-16)
Carbon-hydrogen bond activation reactions of four cycloalkanes (C5H10, C6H12, C7H14, and C8H16) by the Cp'Rh(CO) fragments (Cp' = η(5)-C5H5 (Cp) or η(5)-C5Me5 (Cp*)) were modeled theoretically by combining density functional and coupled cluster theories, and their reaction rates were measured

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service