Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

270202

Sigma-Aldrich

4-Bromophenyl isothiocyanate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4NCS
CAS Number:
Molecular Weight:
214.08
Beilstein:
878549
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
To inquire about this product 270202, please contact your local MilliporeSigma office or dealer. Contact Technical Service

Assay

97%

form

solid

mp

56-58 °C (lit.)

SMILES string

Brc1ccc(cc1)N=C=S

InChI

1S/C7H4BrNS/c8-6-1-3-7(4-2-6)9-5-10/h1-4H

InChI key

XQACWEBGSZBLRG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Bromophenyl isothiocyanate was used in the synthesis of 1-(4-bromophenyl)-3-ethyl-(1,2-dideoxy-D-glycero-α-D-galacto-heptofurano)[2,1-d]imidazolidine-2-thione[1].

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

1-Aryl(glycofurano)imidazolidine-2-thiones derived from new 2-(alkylamino)-2-deoxyheptoses having d-glycero- d-galacto and d-glycero-d-talo configurations.
Galbis Perez JA, et al.
Carbohydrate Research, 131(1), 71-82 (1984)
A Ziegelhöffer et al.
General physiology and biophysics, 2(6), 447-456 (1983-12-01)
Isothiocyanates are potent modifiers of thiol groups, and they have been successfully applied in studying the active site structure of renal (Na+ + K+)-ATPase. However, very little has been known on interactions of isothiocyanates with myocardial sarcolemmal ATPases. In the
J Knopp et al.
Acta endocrinologica, 98(1), 68-72 (1981-09-01)
Liver nuclei bind thyroid hormones (T3 and T4) with different binding affinities. In out studies it was estimated that the apparent association constant (Ka) in the liver nuclei for T3 was 8.8 X 10(10) l mol-1 and for T4 was

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service