Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

269840

Sigma-Aldrich

Tetrahydrofuran-d8

99.5 atom % D

Synonym(s):

THF-d8, Deuterated tetrahydrofuran, Octadeuterotetrahydrofuran

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4D8O
CAS Number:
Molecular Weight:
80.16
Beilstein:
111854
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:

isotopic purity

99.5 atom % D

packaging

pk of 10 × 1 mL

refractive index

n20/D 1.403 (lit.)

bp

65-66 °C (lit.)

mp

−106 °C (lit.)

density

0.985 g/mL at 25 °C (lit.)

mass shift

M+8

SMILES string

[2H]C1([2H])OC([2H])([2H])C([2H])([2H])C1([2H])[2H]

InChI

1S/C4H8O/c1-2-4-5-3-1/h1-4H2/i1D2,2D2,3D2,4D2

InChI key

WYURNTSHIVDZCO-SVYQBANQSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service