Skip to Content
MilliporeSigma
All Photos(2)

Documents

269409

Sigma-Aldrich

2-Nitrophenyl isocyanate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4NCO
CAS Number:
Molecular Weight:
164.12
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

bp

135-137 °C/17 mmHg (lit.)

mp

40-41 °C (lit.)

storage temp.

2-8°C

SMILES string

[O-][N+](=O)c1ccccc1N=C=O

InChI

1S/C7H4N2O3/c10-5-8-6-3-1-2-4-7(6)9(11)12/h1-4H

InChI key

JRVZITODZAQRQM-UHFFFAOYSA-N

Application

2-Nitrophenyl isocyanate was used in the synthesis of symmetrical bis-2-nitrophenylcarbodi-imide via dimerisation reaction.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Intramolecular reaction between nitro and carbodi-imide groups; a new synthesis of 2-arylbenzotriazoles.
Houghton PG, et al.
Journal of the Chemical Society. Perkin Transactions 1, 1471-1479 (1985)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service