Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

268593

Sigma-Aldrich

Cyclopentanepropionyl chloride

98%

Synonym(s):

3-Cyclopentylpropionyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C5H9CH2CH2COCl
CAS Number:
Molecular Weight:
160.64
Beilstein:
1856952
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.464 (lit.)

bp

199-200 °C (lit.)

density

1.049 g/mL at 25 °C (lit.)

functional group

acyl chloride

SMILES string

ClC(=O)CCC1CCCC1

InChI

1S/C8H13ClO/c9-8(10)6-5-7-3-1-2-4-7/h7H,1-6H2

InChI key

SZQVEGOXJYTLLB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Cyclopentanepropionyl chloride was used in the synthesis of 3-cyclopentylpropanamido)methylboronic acid[1].

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

183.2 °F - closed cup

Flash Point(C)

84 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Astrid Zervosen et al.
Bioorganic & medicinal chemistry, 20(12), 3915-3924 (2012-05-15)
In response to the widespread use of β-lactam antibiotics bacteria have evolved drug resistance mechanisms that include the production of resistant Penicillin Binding Proteins (PBPs). Boronic acids are potent β-lactamase inhibitors and have been shown to display some specificity for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service