Skip to Content
MilliporeSigma
All Photos(1)

Documents

268488

Sigma-Aldrich

Niobium

foil, thickness 0.025 mm, 99.8% trace metals basis

Synonym(s):

Columbium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Nb
CAS Number:
Molecular Weight:
92.91
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

99.8% trace metals basis

form

foil

resistivity

13-16 μΩ-cm, 20°C

thickness

0.025 mm

bp

4742 °C (lit.)

mp

2468 °C (lit.)

density

8.57 g/mL at 25 °C (lit.)

SMILES string

[Nb]

InChI

1S/Nb

InChI key

GUCVJGMIXFAOAE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Quantity

4.8 g = 150 × 150 mm

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christos D Malliakas et al.
Journal of the American Chemical Society, 135(5), 1719-1722 (2013-01-23)
2H-NbSe(2) is a canonical Charge-Density-Wave (CDW) layered material the structural details of which remained elusive. We report the detailed structure of 2H-NbSe(2) below the CDW transition using a (3 + 2)-dimensional crystallographic approach on single crystal X-ray diffraction data collected
Yan Sheng et al.
Optics express, 21(4), 4475-4480 (2013-03-14)
We propose and fabricate a LiNbO₃-based nonlinear photonic crystal with locally ordered ferroelectric domains. The nonlinearity modulation provides sets of uniformly distributed reciprocal lattice vectors, ensuring broadband high frequency conversion efficiency. Frequency tripling via cascading is demonstrated in the range
M I Tsindlekht et al.
Journal of physics. Condensed matter : an Institute of Physics journal, 25(8), 085701-085701 (2013-01-31)
We report results of measurements of the ac conductivity of a Nb superconducting thin film in a swept dc magnetic field. In the mixed state the swept dc field creates vortices at the film surface which pass through the film
Kulbir Kaur Ghuman et al.
Journal of physics. Condensed matter : an Institute of Physics journal, 25(8), 085501-085501 (2013-01-25)
A systematic study of electronic structure and band gap states is conducted to analyze the monodoping and charge compensated codoping of rutile TiO(2) with Rh and Nb, using the DFT + U approach. Doping of rutile TiO(2) with Rh atoms induces hybridized
Huaxiang Yi et al.
Optics express, 20(25), 27562-27568 (2012-12-25)
We demonstrate error-free 80km transmission by a silicon carrier-depletion Mach-Zehnder modulator at 10Gbps and the power penalty is as low as 1.15dB. The devices were evaluated through the bit-error-rate characterizations under the system-level analysis. The silicon Mach-Zehnder modulator was also

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service