Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

264555

Sigma-Aldrich

3-Amino-2-methoxydibenzofuran

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H11NO2
CAS Number:
Molecular Weight:
213.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

92-94 °C (lit.)

SMILES string

COc1cc2c(cc1N)oc3ccccc23

InChI

1S/C13H11NO2/c1-15-13-6-9-8-4-2-3-5-11(8)16-12(9)7-10(13)14/h2-7H,14H2,1H3

InChI key

SGQVWNYSHWQNOS-UHFFFAOYSA-N

General description

3-Amino-2-methoxydibenzofuran is an effective inhibitor of 1-nitropyrene metabolism.

Application

3-Amino-2-methoxydibenzofuran has been used in the preparation of end functional poly(N-isopropylacrylamide) (PNIPAM).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P C Howard et al.
Research report (Health Effects Institute), (66)(66), 1-17 (1994-05-01)
Copollutants found in air samples and other complex chemical mixtures may alter the metabolism, and thus the biological activity, of chemical carcinogens. As an initial step to determine whether the metabolism and DNA binding of carcinogenic nitrated polycyclic aromatic hydrocarbons
Urs Rauwald et al.
Chemical communications (Cambridge, England), 47(21), 6000-6002 (2011-04-14)
The chain end complexation of a functional PNIPAM by a cucurbit[8]uril-viologen complex causes a shift in its lower critical solution temperature (LCST) by over 5 °C. An instantaneous phase change of the thermally responsive polymer beyond its LCST can be

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service