Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

264482

Sigma-Aldrich

2,6-Difluorophenylacetic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2C6H3CH2CO2H
CAS Number:
Molecular Weight:
172.13
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

mp

100-102 °C (lit.)

functional group

carboxylic acid
fluoro

SMILES string

OC(=O)Cc1c(F)cccc1F

InChI

1S/C8H6F2O2/c9-6-2-1-3-7(10)5(6)4-8(11)12/h1-3H,4H2,(H,11,12)

InChI key

FUGDCKXBUZFEON-UHFFFAOYSA-N

Application

2,6-Difluorophenylacetic acid has been used in the synthesis of:
  • α-azidoacetophenones[1]
  • N-(2,6-difluorobenzyl)-N′-(1H-5-indazolyl)urea[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masayuki Iwakubo et al.
Bioorganic & medicinal chemistry, 15(1), 350-364 (2006-10-19)
In a previous study, we identified several structurally unrelated scaffolds of the Rho kinase inhibitor using pharmacophore information obtained from the results of a high-throughput screening and structural information from a homology model of Rho kinase. 1H-Indazole is one of
Mariela Bollini et al.
Bioorganic & medicinal chemistry letters, 23(4), 1110-1113 (2013-01-10)
Design of non-nucleoside inhibitors of HIV-1 reverse transcriptase with improved activity towards Tyr181Cys containing variants was pursued with the assistance of free energy perturbation (FEP) calculations. Optimization of the 4-R substituent in 1 led to ethyl and isopropyl analogs 1e

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service