Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

264407

Sigma-Aldrich

3,4-Difluorobenzylamine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2C6H3CH2NH2
CAS Number:
Molecular Weight:
143.13
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.493 (lit.)

density

1.21 g/mL at 25 °C (lit.)

functional group

amine
fluoro

SMILES string

NCc1ccc(F)c(F)c1

InChI

1S/C7H7F2N/c8-6-2-1-5(4-10)3-7(6)9/h1-3H,4,10H2

InChI key

PHLZUDXEBCQHKM-UHFFFAOYSA-N

General description

3,4-Difluorobenzylamine on condensation with glyoxal yields corresponding hexabenzyl substituted hexaazaisowurtzitanes.

Application

3,4-Difluorobenzylamine has been used in the preparation of 2-[(5-chloro-2-methoxyphenyl)sulfonyl(2-fluoroallyl)amino]-N-(3,4-difluorobenzyl)-4-pentenamide.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Polyfluorinated hexabenzyl hexaazaisowurtzitanes.
Kerscher T, et al.
Journal of Fluorine Chemistry, 127(8), 1030-1035 (2006)
A Ring-Closing Metathesis Pathway to Fluorovinyl-Containing Nitrogen Heterocyles.
De Matteis V, et al.
European Journal of Organic Chemistry, 2006(5), 1166-1176 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service