Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

263699

Sigma-Aldrich

3-Ethoxyacrylonitrile, mixture of cis and trans

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OCH=CHCN
CAS Number:
Molecular Weight:
97.12
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.454 (lit.)

bp

90-91 °C/19 mmHg (lit.)

density

0.944 g/mL at 25 °C (lit.)

functional group

ether
nitrile

SMILES string

CCOC=CC#N

InChI

1S/C5H7NO/c1-2-7-5-3-4-6/h3,5H,2H2,1H3

InChI key

HUPVIAINOSTNBJ-UHFFFAOYSA-N

General description

Heck reaction of 3-ethoxyacrylonitrile with aryl bromides catalyzed by a tetraphosphine/palladium complex has been reported.

Application

3-Ethoxyacrylonitrile has been used in the preparation of (Z)-β-aminoacrylonitrile.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heck reactions of 2-substituted enol ethers with aryl bromides catalysed by a tetraphosphine/palladium complex.
Battace A, et al.
Tetrahedron Letters, 47(4), 459-462 (2006)
Sundeep Rayat et al.
Chemical research in toxicology, 18(8), 1211-1218 (2005-08-16)
A discussion of nitrosative deamination of cytosine 1 is presented that argues for the formation of 6 by diazotization of 1 to cytosinediazonium ion 2 and its electrostatic complex 3, dediazoniation to 4 <--> 5, and amide-bond cleavage to 6.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service