Skip to Content
MilliporeSigma
All Photos(1)

Documents

263443

Sigma-Aldrich

1-Bromo-2,4-difluorobenzene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H3F2
CAS Number:
Molecular Weight:
192.99
Beilstein:
1680892
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.505 (lit.)

bp

145-146 °C (lit.)

mp

−4 °C (lit.)

density

1.708 g/mL at 25 °C (lit.)

SMILES string

Fc1ccc(Br)c(F)c1

InChI

1S/C6H3BrF2/c7-5-2-1-4(8)3-6(5)9/h1-3H

InChI key

MGHBDQZXPCTTIH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Bromo-2,4-difluorobenzene undergoes lithiation exclusively at the position having two adjacent halogen substituents, to yield 6-bromo-2,3-difluorobenzoic acid.

Application

1-Bromo-2,4-difluorobenzene has been used in:
  • preparation of (2S)-1-(2,4-difluorophenyl)-2-(1,1-dimethyl-1-sila-ethoxy)-propan-1-one
  • enantiomeric preparation of the key intermediate of chiral azole antifungal agents by a chemoenzymatic process

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

125.6 °F - closed cup

Flash Point(C)

52 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ram Shankar Upadhayaya et al.
Bioorganic & medicinal chemistry, 12(9), 2225-2238 (2004-04-15)
A series of (2R,3S)-2-(2,4-difluorophenyl)-3-(5-[2-[4-aryl-piperazin-1-yl]-ethyl]-tetrazol-2-yl)-1-[1,2,4]-triazol-1-yl-butan-2-ol (11a-n) and (2R,3S)-2-(2,4-difluorophenyl)-3-(5-[2-[4-aryl-piperazin-1-yl]-ethyl]-tetrazole-1-yl)-1-[1,2,4]-triazol-1-yl-butan-2-ol (12a-n) has been synthesized. The antifungal activity of compounds was evaluated by in vitro agar diffusion and broth dilution assay. Compounds 11d and its positional isomer 12d having 3-trifluoromethyl substitution on the phenyl
A practical chemoenzymatic synthesis of a key intermediate of antifungal agents.
Yasohara Y, et al.
Tetrahedron Letters, 42(19), 3331-3333 (2001)
Regioselective ortho-lithiation of chloro and bromo substituted fluoroarenes.
Mongin F and Schlosser M.
Tetrahedron Letters, 37(36), 6551-6554 (1996)
Ji Gwang Yu et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 23(63), 16044-16050 (2017-08-24)
Four dibenzofuran-type host materials substituted with a carbazolylcarbazole moiety were synthesized to investigate the effect of substitution position on the material parameters and device performances of host materials. The carbazolylcarbazole moiety was substituted at the 1-, 2-, 3-, and 4-positions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service