Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

263311

Sigma-Aldrich

Tellurium

ingot, 99.999% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Te
CAS Number:
Molecular Weight:
127.60
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

99.999% trace metals basis

form

ingot

resistivity

5.8-33 μΩ-cm, 20°C

bp

990 °C (lit.)

mp

450 °C (lit.)

density

6.24 g/mL at 25 °C (lit.)

SMILES string

[Te]

InChI

1S/Te

InChI key

PORWMNRCUJJQNO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 4 - Repr. 1B - Skin Sens. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vijay P Singh et al.
Organic letters, 15(24), 6274-6277 (2013-11-28)
In an effort to improve the chain-breaking capacity of the natural antioxidants, an octyltelluro group was introduced next to the phenolic moiety in β- and δ-tocopherol. The new vitamin E analogues quenched peroxyl radicals more efficiently than α-tocopherol and were
Raymond J Turner et al.
Biotechnology advances, 30(5), 954-963 (2011-09-13)
Here, we overview the most recent advances in understanding the bacterial mechanisms that stay behind the reduction of tellurium oxyanions in both planktonic cells and biofilms. This is a topic of interest for basic and applied research because microorganisms are
Biomethylation of selenium and tellurium: microorganisms and plants.
Thomas G Chasteen et al.
Chemical reviews, 103(1), 1-25 (2003-01-09)
Zhe Liu et al.
Optics express, 21(6), 7799-7810 (2013-04-03)
ZnTe is an important p-type semiconductor with great applications as field-effect transistors and photodetectors. In this paper, individual ZnTe nanowires based field-effect transistors was fabricated, showing evident p-type conductivity with an effect mobility of 11.3 cm(2)/Vs. Single ZnTe nanowire based
Guizhong Wang et al.
Optics express, 21(4), 4703-4708 (2013-03-14)
The octave-spanning spectrum was generated in a tellurite glass based microstructured fiber pumped by a 528 MHz repetition rate Yb:fiber ring laser without amplification. The laser achieved 40% output optical-to-optical efficiency with the output power of 410 mW. By adjusting

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service