Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

262692

Sigma-Aldrich

Phenanthridine

≥99%, purified by sublimation

Synonym(s):

Benzo[c]quinoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H9N
CAS Number:
Molecular Weight:
179.22
Beilstein:
120204
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

≥99%

form

solid

purified by

sublimation

bp

349 °C/769 mmHg (lit.)

mp

104-107 °C (lit.)

SMILES string

c1ccc2c(c1)cnc3ccccc23

InChI

1S/C13H9N/c1-2-6-11-10(5-1)9-14-13-8-4-3-7-12(11)13/h1-9H

InChI key

RDOWQLZANAYVLL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Harrie J M Gijsen et al.
Journal of medicinal chemistry, 53(19), 7011-7020 (2010-09-03)
The TRPA1 channel can be considered as a key biological sensor to irritant chemicals. In this paper, the discovery of 11H-dibenz[b,e]azepines (morphanthridines) and dibenz[b,f][1,4]oxazepines is described as extremely potent agonists of the TRPA1 receptor. This has led to the discovery
Alfonso Pérez-Garrido et al.
Bioorganic & medicinal chemistry, 17(2), 896-904 (2008-12-06)
This paper reports a QSAR study for predicting the complexation of a large and heterogeneous variety of substances (233 organic compounds) with beta-cyclodextrins (beta-CDs). Several different theoretical molecular descriptors, calculated solely from the molecular structure of the compounds under investigation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service