Skip to Content
MilliporeSigma
All Photos(1)

Documents

261785

Sigma-Aldrich

Barium diphenylamine-4-sulfonate

98%

Synonym(s):

4-(Phenylamino)benzenesulfonic acid barium salt, Diphenylamine-4-sulfonic acid barium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C12H10NO3S)2Ba
CAS Number:
Molecular Weight:
633.88
Beilstein:
3795146
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

98%

SMILES string

[Ba++].[O-]S(=O)(=O)c1ccc(Nc2ccccc2)cc1.[O-]S(=O)(=O)c3ccc(Nc4ccccc4)cc3

InChI

1S/2C12H11NO3S.Ba/c2*14-17(15,16)12-8-6-11(7-9-12)13-10-4-2-1-3-5-10;/h2*1-9,13H,(H,14,15,16);/q;;+2/p-2

InChI key

IVCNVXFNTKXMCA-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service