Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

261742

Sigma-Aldrich

N-Benzyl-2-phenethylamine

99%

Synonym(s):

N-Benzyl-2-phenylethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH2NHCH2C6H5
CAS Number:
Molecular Weight:
211.30
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

refractive index

n20/D 1.566 (lit.)

bp

327-328 °C/750 mmHg (lit.)

density

1.007 g/mL at 25 °C (lit.)

SMILES string

C(Cc1ccccc1)NCc2ccccc2

InChI

1S/C15H17N/c1-3-7-14(8-4-1)11-12-16-13-15-9-5-2-6-10-15/h1-10,16H,11-13H2

InChI key

UPABQMWFWCMOFV-UHFFFAOYSA-N

Application

N-Benzyl-2-phenethylamine has been used in synthesis of dithiocarbamates.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elton Luiz Silva et al.
Journal of nanoscience and nanotechnology, 16(2), 1291-1300 (2016-07-20)
All-trans retinoic acid, a hydrophobic drug, has become one of the most successful examples of differentiation agents used for treatment of acute promyelocytic leukemia. On the other hand, histone deacetylase inhibitors, such as cholesteryl butyrate, present differentiating activity and.can potentiate
N Segovia et al.
Journal of applied toxicology : JAT, 22(6), 353-357 (2002-11-09)
Dithiocarbamates (DTCs) are chemicals featuring a great chelating capacity. The toxicological study of DTCs is very important in view of their relatively simple synthesis and wide array of sanitary and industrial applications. In this study, the toxicity of some of
Zsanett Dorkó et al.
Molecules (Basel, Switzerland), 23(6) (2018-05-31)
One of the main reasons for making molecularly imprinted polymers (MIPs) has been that MIPs interact selectively with a specific target compound. This claim is investigated here with the example of a widely used type of noncovalent MIP, the MIP
Valquíria T Arantes et al.
Colloids and surfaces. B, Biointerfaces, 188, 110749-110749 (2020-01-14)
Repair of tissue damaged in diabetic wounds is essential to minimize the cases of amputation of the limbs in millions of diabetic people around the world. Although the all-trans retinoic acid (ATRA) is described as a potential wound healing agent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service