Skip to Content
MilliporeSigma
All Photos(1)

Documents

26130

Sigma-Aldrich

3-Chloropropionaldehyde diethylacetal

technical, ≥90% (GC)

Synonym(s):

3-Chloro-1,1-diethoxypropane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CH2CH(OC2H5)2
CAS Number:
Molecular Weight:
166.65
Beilstein:
1698609
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Quality Level

Assay

≥90% (GC)

contains

~0.5% CaCO3 as stabilizer

refractive index

n20/D 1.42 (lit.)
n20/D 1.420

bp

84 °C/25 mmHg (lit.)

density

0.995 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCOC(CCCl)OCC

InChI

1S/C7H15ClO2/c1-3-9-7(5-6-8)10-4-2/h7H,3-6H2,1-2H3

InChI key

NXHONHDWVLPPCS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Chloropropionaldehyde diethylacetal has been used in the synthesis of:
  • N-[4-[[3-(2, 4-diamino-1,6-dihydro-6-oxo-5-pyrimidinyl)propyl]amino]benzoyl]-L-glutamic acid, an acyclic analog of 5,6,7,8-tetrahydrofolic acid
  • 1-(D,L)-(3′-amino, 3′-carboxypropyl)uracil, nucleoamino acid
  • 7-methoxychromanone
  • grignard reagent

Other Notes

Masked 3-chloropropionaldehyde; preparation of Grignard reagent

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

129.2 °F - closed cup

Flash Point(C)

54 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An Efficient Preparation of 7-Methoxychromanone.
Cao Z and Liehr JG.
Synthetic Communications, 26(6), 1233-1245 (1996)
Masoomeh Shirzad et al.
Assay and drug development technologies, 17(5), 231-239 (2019-05-14)
This study aimed to synthesize methoxy polyethylene glycol propionaldehyde (mPEG20,000-ALD) for the preparation of PEGylated nanoliposomal cisplatin. Nanocarriers such as liposomes are developed for a wide range of drug delivery systems. PEG with high molecular weight (Mw) is used to
J L Kelley et al.
Journal of medicinal chemistry, 33(2), 561-567 (1990-02-01)
The synthesis and biological evaluation of N-[4-[[3-(2,4-diamino-1,6-dihydro-6-oxo-5-pyrimidinyl)propyl]amino]- benzoyl]-L-glutamic acid (1) (5-DACTHF, 543U76), an acyclic analogue of 5,6,7,8-tetrahydrofolic acid (THFA), are described. The key intermediate, hemiaminal 8, was prepared in four stages from 3-chloropropionaldehyde diethyl acetal. Reaction of 8 with dimethyl
A. Greiner
Tetrahedron Letters, 30, 3547-3547 (1989)
A B Cheikh et al.
Journal of molecular evolution, 30(4), 315-321 (1990-04-01)
We have prepared the nucleoamino acids 1-(3'-amino,3'-carboxypropyl)uracil (3) and 9-(3'-amino,3'-carboxypropyl)adenine (4) as (L)-enantiomers and as racemic mixtures. When 3 or 4 is suspended in water and treated with N,N'-carbonyldiimidazole, peptides are formed in good yield. The products formed from the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service