Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

260576

Sigma-Aldrich

2,3,4-Trihydroxybenzophenone

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)3C6H2COC6H5
CAS Number:
Molecular Weight:
230.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

139-141 °C (lit.)

solubility

ethanol: soluble 2%, clear, yellow to very dark yellow-orange

functional group

ketone
phenyl

SMILES string

Oc1ccc(c(O)c1O)C(=O)c2ccccc2

InChI

1S/C13H10O4/c14-10-7-6-9(12(16)13(10)17)11(15)8-4-2-1-3-5-8/h1-7,14,16-17H

InChI key

HTQNYBBTZSBWKL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,3,4-Trihydroxybenzophenone is a hydroxylated benzophenone UV filter and has been quantitated in sea water samples by dispersive liquid-liquid microextraction followed by GC-MS. Estrogenic activity of 2,3,4-trihydroxybenzophenone has been tested by yeast-two hybrid assay.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Estrogenicity of metabolites of benzophenone derivatives examined by a yeast two-hybrid assay.
Takatori S, et al.
Journal of Health Science, 49(2), 91-98 (2003)
Isuha Tarazona et al.
Journal of chromatography. A, 1217(29), 4771-4778 (2010-06-19)
A new analytical method for the determination of four hydroxylated benzophenone UV filters (i.e. 2-hydroxy-4-methoxybenzophenone (HMB), 2,4-dihydroxybenzophenone (DHB), 2,2'-dihydroxy-4-methoxybenzophenone (DHMB) and 2,3,4-trihydroxybenzophenone (THB)) in sea water samples is presented. The method is based on dispersive liquid-liquid microextraction (DLLME) followed by
C A Downs et al.
Archives of environmental contamination and toxicology, 70(2), 265-288 (2015-10-22)
Toxicity persistence to the nontarget amphipod Hyalella curvispina in runoff events following chlorpyrifos applications to soy experimental plots was compared in conventional and no-till management. Two application scenarios were compared: an early-season application with the soil almost bare and a
Yoko Watanabe et al.
Toxicology and applied pharmacology, 282(2), 119-128 (2014-12-22)
Benzophenone-3 (2-hydroxy-4-methoxybenzophenone; BP-3) is widely used as sunscreen for protection of human skin and hair from damage by ultraviolet (UV) radiation. In this study, we examined the metabolism of BP-3 by rat and human liver microsomes, and the estrogenic and
S C Cunha et al.
Environmental research, 143(Pt B), 65-71 (2015-05-20)
In the last decades, awareness regarding personal care products (PCP), i.e. synthetic organic chemicals frequently used in cosmetic and hygienic products, has become a forward-looking issue, due to their persistency in the environment and their potential multi-organ toxicity in both

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service