Skip to Content
MilliporeSigma
All Photos(1)

Documents

259837

Sigma-Aldrich

cis-11-Hexadecen-1-yl acetate

95%

Synonym(s):

Z-11-HDA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H34O2
CAS Number:
Molecular Weight:
282.46
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

95%

SMILES string

CCCC\C=C/CCCCCCCCCCOC(C)=O

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

141.8 °F - closed cup

Flash Point(C)

61 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service