Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

25970

Sigma-Aldrich

3-Chlorophenylhydrazine hydrochloride

≥97.0% (AT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4NHNH2 · HCl
CAS Number:
Molecular Weight:
179.05
Beilstein:
3565828
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥97.0% (AT)

form

solid

mp

240-245 °C (dec.) (lit.)

SMILES string

Cl[H].NNc1cccc(Cl)c1

InChI

1S/C6H7ClN2.ClH/c7-5-2-1-3-6(4-5)9-8;/h1-4,9H,8H2;1H

InChI key

CRRIAWUJYMLJOE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Chlorophenylhydrazine hydrochloride has been used in the preparation of:
  • 1-(3-chlorophenyl)-3,5-diphenyl-1H-pyrazolo[4,3-c]quinolin-4(5H)-one[1]
  • 11-(3-chlorophenyl)-9-phenyl-5,6-dihydro-4H,11H-benzo[i,j]- pyrazolo[3,4-b]quinolizin-8-one[1]
  • 1-(3-chlorophenyl)-3-(cyclohexyl)-5-(4-phenoxy phenyl)pyrazole[2]

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Akihiko Tanitame et al.
Bioorganic & medicinal chemistry, 12(21), 5515-5524 (2004-10-07)
The 4-piperidyl moiety and the pyrazole ring in 1-(3-chlorophenyl)-5-(4-phenoxyphenyl)-3-(4-piperidyl)pyrazole 2, which has previously shown improved DNA gyrase inhibition and target-related antibacterial activity, were transformed to other groups and the in vitro antibacterial activity of the synthesized compounds was evaluated. The
Ring closure reactions of 3-arylhydrazonoalkyl-quinolin-2-ones to 1-aryl-pyrazolo [4, 3-c] quinolin-2-ones.
Stadlbauer W and Hojas G.
Journal of Heterocyclic Chemistry, 41(5), 681-690 (2004)
Amina Othmani et al.
Environmental science and pollution research international, 26(25), 25969-25984 (2019-07-06)
This paper suggests a new alternative for the acceleration of dye removal by adopting alternating current instead of direct current in the treatment of methylene blue solutions and industrials effluents, using anodic oxidation on Pb/PbO2 and stainless steel (SS)/PbO2 anodes.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service