Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

258970

Sigma-Aldrich

2,4,5-Trifluoronitrobenzene

99%

Synonym(s):

1,2,4-Trifluoro-5-nitrobenzene, 5-Nitro-1,2,4-trifluorobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F3C6H2NO2
CAS Number:
Molecular Weight:
177.08
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.4943 (lit.)

bp

194-195 °C (lit.)

mp

−11 °C (lit.)

density

1.544 g/mL at 25 °C (lit.)

functional group

fluoro
nitro

SMILES string

[O-][N+](=O)c1cc(F)c(F)cc1F

InChI

1S/C6H2F3NO2/c7-3-1-5(9)6(10(11)12)2-4(3)8/h1-2H

InChI key

ROJNMGYMBLNTPK-UHFFFAOYSA-N

Application

2,4,5-Trifluoronitrobenzene may be used in chemical synthesis.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

192.2 °F - closed cup

Flash Point(C)

89 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service