Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

258261

Sigma-Aldrich

Chlorophyll b from spinach

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C55H70MgN4O6
CAS Number:
Molecular Weight:
907.47
Beilstein:
4122778
EC Number:
MDL number:
UNSPSC Code:
12352100

mp

183-185 °C (lit.)

λmax

643 nm

SMILES string

[H]C(=O)c1c(CC)c2cc3c(C)c4C(=O)[C@H](C(=O)OC)c5c6nc(cc7c(C)c(C=C)c(cc1n2)n7[Mg]n3c45)[C@@H](C)[C@@H]6CCC(=O)OC\C=C(/C)CCC[C@H](C)CCC[C@H](C)CCCC(C)C

InChI

1S/C55H71N4O6.Mg/c1-12-38-35(8)42-27-43-36(9)40(23-24-48(61)65-26-25-34(7)22-16-21-33(6)20-15-19-32(5)18-14-17-31(3)4)52(58-43)50-51(55(63)64-11)54(62)49-37(10)44(59-53(49)50)28-46-39(13-2)41(30-60)47(57-46)29-45(38)56-42;/h12,25,27-33,36,40,51H,1,13-24,26H2,2-11H3,(H-,56,57,58,59,60,62);/q-1;+2/p-1/b34-25+;/t32-,33-,36+,40+,51-;/m1./s1

InChI key

NSMUHPMZFPKNMZ-VBYMZDBQSA-M

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Chlorophyll b extends the absorption spectrum of light-harvesting complexes LHC-I and LHC-II above 700 nm, a region of the visible spectrum that is not absorbed when only chlorophyll a is present in the LHCs.

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service