Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

257966

Sigma-Aldrich

2,5-Dimethoxybenzoic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2C6H3CO2H
CAS Number:
Molecular Weight:
182.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

76-78 °C (lit.)

SMILES string

COc1ccc(OC)c(c1)C(O)=O

InChI

1S/C9H10O4/c1-12-6-3-4-8(13-2)7(5-6)9(10)11/h3-5H,1-2H3,(H,10,11)

InChI key

NYJBTJMNTNCTCP-UHFFFAOYSA-N

Application

2,5-Dimethoxybenzoic acid has been used in synthesis of:
  • galbulimima alkaloid GB 13
  • 3,4-dihydrofluoren-2(1H)-ones via reductive alkylation

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lewis N Mander et al.
Journal of the American Chemical Society, 125(9), 2400-2401 (2003-02-27)
This contribution describes a synthetic approach to alkaloid GB 13, previously isolated from the North Australian and Papua New Guinean rain forest tree Galbulimima belgraveana. A Birch reductive alkylation of 2,5-dimethoxybenzoic acid by 3-methoxybenzyl bromide, followed by an acid-catalyzed cyclization
Reductive alkylation of 2, 5-dimethoxybenzoic acid: a direct synthesis of dihydrofluoren-2-ones.
Hook JM and Mander LN.
The Journal of Organic Chemistry, 45(9), 1722-1724 (1980)
Erick F de Oliveira et al.
Journal of photochemistry and photobiology. B, Biology, 214, 112081-112081 (2020-11-27)
The objective of this study was to investigate synergistic antibacterial activity based on a combination of UV-A light and three classes of food grade compounds: benzoic acid derivatives, cinnamic acid derivatives, and gallates. By using Escherichia coli O157:H7 as the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service