Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

257885

Sigma-Aldrich

Trimethylsilyl cyclopentadiene, mixture of isomers

Synonym(s):

NSC 176274

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$507.00

$507.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$507.00

About This Item

Empirical Formula (Hill Notation):
C8H14Si
CAS Number:
Molecular Weight:
138.28
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$507.00


Please contact Customer Service for Availability

Request a Bulk Order

form

liquid

refractive index

n20/D 1.471 (lit.)

bp

138-140 °C (lit.)

density

0.833 g/mL at 25 °C (lit.)

storage temp.

−20°C

SMILES string

C[Si](C)(C)C1C=CC=C1

InChI

1S/C8H14Si/c1-9(2,3)8-6-4-5-7-8/h4-8H,1-3H3

InChI key

VMFHCJPMKUTMMQ-UHFFFAOYSA-N

General description

Trimethylsilyl cyclopentadiene reacts with ruthenium trichloride in alcohol solvent to yield ruthenocene[1].

Application

Reactant involved in:
  • Diels-Alder reactions
  • Synthesis of ring-functionalized molybdenocene complexes
  • Allyl isomerization
  • Aromatization of C5 rings
  • Synthesis of N-hydroxyindoles via nucleophilic addition

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

84.2 °F - closed cup

Flash Point(C)

29 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multiple desilylation of silylated cyclopentadienes promoted by ionic ruthenium and rhodium halides.
Winter CH, et al.
Inorganic Chemistry, 32(17), 3654-3659 (1993)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service