Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

257842

Sigma-Aldrich

Perfluorodecyl iodide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3(CF2)9I
CAS Number:
Molecular Weight:
645.98
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

bp

195-200 °C (lit.)

mp

65-67 °C (lit.)

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)I

InChI

1S/C10F21I/c11-1(12,3(15,16)5(19,20)7(23,24)9(27,28)29)2(13,14)4(17,18)6(21,22)8(25,26)10(30,31)32

InChI key

UDWBMXSQHOHKOI-UHFFFAOYSA-N

Application

Perfluorodecyl iodide has been used in preparation of:
  • 3,5-bis(perfluorodecyl)phenylboronic acid, "green" catalyst for the direct amide condensation reaction
  • 3-(perfluorodecyl)prop-1-ene

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

3, 5-Bis (perfluorodecyl) phenylboronic acid as an easily recyclable direct amide condensation catalyst.
Ishihara K, et al.
Synlett, 2001(9), 1371-1374 (2001)
Synthesis of perfluorinated allylic compounds by radical allylation and their purification over fluorous reverse-phase silica.
Ryu I, et al.
Tetrahedron Letters, 42(5), 947-950 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service