Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

257044

Sigma-Aldrich

L-Selectride® solution

1.0 M lithium tri-sec-butylborohydride in THF

Synonym(s):

Lithium tri-sec-butylborohydride solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Li[CH(CH3)CH2CH3]3BH
CAS Number:
Molecular Weight:
190.10
Beilstein:
4007327
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

form

liquid

Quality Level

reaction suitability

reagent type: reductant

concentration

1.0 M lithium tri-sec-butylborohydride in THF

density

0.89 g/mL at 25 °C

SMILES string

[Li+].[H][B-](C(C)CC)(C(C)CC)C(C)CC

InChI

1S/C12H28B.Li/c1-7-10(4)13(11(5)8-2)12(6)9-3;/h10-13H,7-9H2,1-6H3;/q-1;+1

InChI key

ACJKNTZKEFMEAK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Reducing agent

Reagent for:
  • Enantioselective synthesis of a-amino acids via reduction of N-tert-butanesulfinyl ketimine esters
  • Diastereoselective reduction reactions
  • Hydride reduction of the Danishefsky pyranones
  • Asymmetric reductive aldol reaction of enones with a-alkoxy aldehydes
  • Synthesis of alkanols by carbonylation reactions

Packaging

Supplied in a Sure/Pac cylinder and has a 1/4-turn bronze ball valve with a female 1/4" NPT outlet thread (Z122661) installed. Before using the cylinder, ensure that the valve is closed, then remove carbon steel plug that seals the outlet valve.

Compatible with the following:For instructions please refer to technical bulletin AL-136: The Aldrich Sure/Pac Cylinder System

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Selectride is a registered trademark of Merck KGaA, Darmstadt, Germany
Sure/Pac is a trademark of Sigma-Aldrich Co. LLC

also commonly purchased with this product

Product No.
Description
Pricing

septum inlet adapter

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Carc. 2 - Flam. Liq. 2 - Pyr. Liq. 1 - Skin Corr. 1B - STOT SE 3 - Water-react 1

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service