Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

257028

Sigma-Aldrich

Isopropylmagnesium chloride solution

2.0 M in diethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHMgCl
CAS Number:
Molecular Weight:
102.85
Beilstein:
4124221
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

concentration

2.0 M in diethyl ether

density

0.834 g/mL at 25 °C

SMILES string

CC(C)[Mg]Cl

InChI

1S/C3H7.ClH.Mg/c1-3-2;;/h3H,1-2H3;1H;/q;;+1/p-1

InChI key

GPIBKUJXKCEZOH-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Recommended products

Angled septum-inlet adapter Z118206 or straight septum-inlet adapter, 6mm I.D. inlet, Z118141 or straight septum-inlet adapter, 13mm I.D. inlet, Z118192 is recommended.

Legal Information

Sure/Pac is a trademark of Sigma-Aldrich Co. LLC

septum inlet adapter

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3 - Water-react 1

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

-40.0 °F - closed cup

Flash Point(C)

-40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service