Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

256498

Sigma-Aldrich

Zinc cyanide

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Zn(CN)2
CAS Number:
Molecular Weight:
117.42
Beilstein:
4124366
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Assay:
98%
form:
powder

Assay

98%

form

powder

technique(s)

HPLC: suitable

density

1.85 g/mL at 25 °C (lit.)

SMILES string

N#C[Zn]C#N

InChI

1S/2CN.Zn/c2*1-2;

InChI key

VJHHEOJUJAQMPF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Colloidal quantum dots (CQDs) are semiconducting crystals of only a few nanometers (ca. 2–12 nm) coated with ligand/surfactant molecules to help prevent agglomeration.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service