Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

25625

Sigma-Aldrich

2-Chloro-5-nitrotoluene

≥99.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6ClNO2
CAS Number:
Molecular Weight:
171.58
Beilstein:
2044766
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0% (GC)

form

solid

mp

40-44 °C

SMILES string

Cc1cc(ccc1Cl)[N+]([O-])=O

InChI

1S/C7H6ClNO2/c1-5-4-6(9(10)11)2-3-7(5)8/h2-4H,1H3

InChI key

BGDCQZFFNFXYQC-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phase transfer catalysis used in synthesis of N-(2-methyl-4-nitrophenyl) piperidine [J].
Wenqi YSXXL.
Chemical Reagents, 1, 013-013 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service