Skip to Content
MilliporeSigma
All Photos(1)

Documents

255939

Sigma-Aldrich

Phosphorus, red

lumps, 99.999% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
P
CAS Number:
Molecular Weight:
30.97
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

vapor density

0.02 (vs air)

vapor pressure

0.03 mmHg ( 21 °C)
1 mmHg ( 76.6 °C)

Assay

99.999% trace metals basis

form

lumps

autoignition temp.

500 °F

resistivity

10 μΩ-cm, 20°C

mp

416 °C (subl.) (lit.)

density

2.34 g/mL at 25 °C (lit.)

SMILES string

[P]

InChI

1S/P

InChI key

OAICVXFJPJFONN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manuel Delgado-Baquerizo et al.
Nature, 502(7473), 672-676 (2013-11-01)
The biogeochemical cycles of carbon (C), nitrogen (N) and phosphorus (P) are interlinked by primary production, respiration and decomposition in terrestrial ecosystems. It has been suggested that the C, N and P cycles could become uncoupled under rapid climate change
Yuki Fujita et al.
Nature, 505(7481), 82-86 (2013-11-19)
Plant species diversity in Eurasian wetlands and grasslands depends not only on productivity but also on the relative availability of nutrients, particularly of nitrogen and phosphorus. Here we show that the impacts of nitrogen:phosphorus stoichiometry on plant species richness can
Jacques C Finlay et al.
Science (New York, N.Y.), 342(6155), 247-250 (2013-10-12)
Human activities have increased the availability of reactive nitrogen in many ecosystems, leading to negative impacts on human health, biodiversity, and water quality. Freshwater ecosystems, including lakes, streams, and wetlands, are a large global sink for reactive nitrogen, but factors
Bon-Chul Koo et al.
Science (New York, N.Y.), 342(6164), 1346-1348 (2013-12-18)
Phosphorus ((31)P), which is essential for life, is thought to be synthesized in massive stars and dispersed into interstellar space when these stars explode as supernovae (SNe). Here, we report on near-infrared spectroscopic observations of the young SN remnant Cassiopeia
Zhiguo Cao et al.
Environment international, 65, 100-106 (2014-02-01)
This study documents the temporal variability in concentrations of flame retardants (FRs) in floor dust from three offices in Beijing, China. Dust from Office A (OAD) was collected weekly from March to August, 2012, and sampling of dust from Office

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service