Skip to Content
MilliporeSigma
All Photos(1)

Documents

255491

Sigma-Aldrich

4-Chloromethylstilbene, predominantly trans

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=CHC6H4CH2Cl
CAS Number:
Molecular Weight:
228.72
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

mp

113-117 °C (lit.)

SMILES string

ClCc1ccc(cc1)\C=C\c2ccccc2

InChI

1S/C15H13Cl/c16-12-15-10-8-14(9-11-15)7-6-13-4-2-1-3-5-13/h1-11H,12H2/b7-6+

InChI key

IWJQBYQAGGHNAB-VOTSOKGWSA-N

General description

Generation of highly basic stilbene dianions of 4-chloromethylstilbene by electrochemical reduction in ionic liquids at room temperature has been reported.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Omar Abdul-Rahim et al.
Analytical chemistry, 85(12), 6113-6120 (2013-05-11)
Three highly aprotic bis(trifluoromethylsulfonyl)amide (NTf2(-)) based ionic liquids (ILs) containing the cations trihexyl(tetradecyl)phosphonium (P6,6,6,14(+)), N-butyl-N-methylpyrrolidinium (Pyrr4,1(+)), and (trimethylamine)(dimethylethylammine)dihydroborate ((N111)(N112)BH2(+)) have been examined as media for room temperature voltammetric detection of highly basic stilbene dianions electrochemically generated by the reduction of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service