Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

255114

Sigma-Aldrich

3-Nitroanisole

99%

Synonym(s):

1-Methoxy-3-nitrobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4OCH3
CAS Number:
Molecular Weight:
153.14
Beilstein:
1865326
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

bp

121-123 °C/8 mmHg (lit.)

mp

36-38 °C (lit.)

solubility

alcohol: soluble(lit.)
water: insoluble(lit.)

SMILES string

COc1cccc(c1)[N+]([O-])=O

InChI

1S/C7H7NO3/c1-11-7-4-2-3-6(5-7)8(9)10/h2-5H,1H3

InChI key

WGYFINWERLNPHR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Photoinduced nucleophilic substitution reactions of 3-nitroanisole in the presence of OH in mixtures of H2O and CH3CN and in the presence of OCH3 in CH3OH were investigated by nanosecond time-resolved absorption spectroscopy. 3-Nitroanisole undergoes photoreaction with n-butylamine to yield 3-nitrophenol. It forms a 1:1 inclusion complex with β-cyclodextrin in aqueous buffer solution at pH 12.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spectroscopic and kinetic study of the photoinduced methoxy substitution of 3-nitroanisole and 3, 5-dinitroanisole.
Van Zeijl PHM, et al.
Journal of Photochemistry, 29(3), 415-433 (1985)
Modification of an intermolecular photoreaction by cyclodextrin: the photohydroxylation of 3-nitroanisole.
Evans CH and Gunnlaugsson T.
Journal of Photochemistry and Photobiology A: Chemistry, 78(1), 57-62 (1994)
Nitrophenyl ethers as possible photoaffinity labels. The nucleophilic aromatic photosubstitution revisited.
Cervelo J., et al.
Tetrahedron Letters, 25(37), 4147-4150 (1984)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service