Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

254665

Sigma-Aldrich

3-Nitrobenzenesulfonyl chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4SO2Cl
CAS Number:
Molecular Weight:
221.62
Beilstein:
746542
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

61-62 °C (lit.)

functional group

nitro

SMILES string

[O-][N+](=O)c1cccc(c1)S(Cl)(=O)=O

InChI

1S/C6H4ClNO4S/c7-13(11,12)6-3-1-2-5(4-6)8(9)10/h1-4H

InChI key

MWWNNNAOGWPTQY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Nitrobenzenesulfonyl chloride has been used in preparation of:
  • acyl-2-aminobenzimidazole analogs[1]
  • 6-chloro-2-methyl-3-{1-[(3-nitrophenyl)sulfonyl]-1H-pyrazol-3-yl}imidazo[1,2-a]pyridine[2]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masahiko Hayakawa et al.
Bioorganic & medicinal chemistry, 15(1), 403-412 (2006-10-20)
3-{1-[(4-Fluorophenyl)sulfonyl]-1H-pyrazol-3-yl}-2-methylimidazo[1,2-a]pyridine, 2a, was discovered in our chemical library as a novel p110alpha inhibitor with an IC(50) of 0.67microM, through screening in a scintillation proximity assay. Optimization of the substituents of 2a increased the p110alpha inhibitory activity by more than 300-fold
Jay P Powers et al.
Bioorganic & medicinal chemistry letters, 16(11), 2842-2845 (2006-03-28)
High-throughput screening of a small-molecule compound library resulted in the identification of a novel series of N-acyl 2-aminobenzimidazoles that are potent inhibitors of interleukin-1 receptor-associated kinase-4.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service