Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

253022

Sigma-Aldrich

(S)-(+)-O-Acetylmandelic acid

99%

Synonym(s):

(S)-(+)-α-Acetoxyphenylacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2CH(C6H5)CO2H
CAS Number:
Molecular Weight:
194.18
Beilstein:
2694109
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

optical activity

[α]28/D +153°, c = 2 in acetone

optical purity

ee: 98% (GLC)

mp

97-99 °C (lit.)

functional group

carboxylic acid
ester
phenyl

SMILES string

CC(=O)O[C@H](C(O)=O)c1ccccc1

InChI

1S/C10H10O4/c1-7(11)14-9(10(12)13)8-5-3-2-4-6-8/h2-6,9H,1H3,(H,12,13)/t9-/m0/s1

InChI key

OBCUSTCTKLTMBX-VIFPVBQESA-N

Application

The (R)- and (S)-isomers are chiral derivatizing agents for NMR determination of enantiomeric purity of α-deuterated carboxylic acids, alcohols, and amines.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

J. Chem. Soc. Perkin Trans. II, 83-83 (1983)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service