Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

252654

Sigma-Aldrich

N-(Diphenylmethylene)aminoacetonitrile

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2C=NCH2CN
CAS Number:
Molecular Weight:
220.27
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

98%

mp

84-85 °C (lit.)

solubility

95% ethanol: soluble 50 mg/mL, clear, colorless to faintly yellow

storage temp.

2-8°C

SMILES string

N#CC\N=C(\c1ccccc1)c2ccccc2

InChI

1S/C15H12N2/c16-11-12-17-15(13-7-3-1-4-8-13)14-9-5-2-6-10-14/h1-10H,12H2

InChI key

VRLJFRODHVSTIK-UHFFFAOYSA-N

General description

N-(Diphenylmethylene)aminoacetonitrile is a synthon for a variety of amino acids via phase-transfer-catalyzed alkylation and hydrolysis.[1]

Application

N-(Diphenylmethylene)aminoacetonitrile has been used in:
  • synthesis of ring-substituted phenylalanine amides[2] and o-carboranylalanine, a potential analog of phenylalanine[3]
  • a key enantiodifferentiating step in concise synthesis of dehydrocoronamic acid ethyl ester[4]
  • synthesis of γ-amino acid esters[5]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A general, convenient way to carborane-containing amino acids for boron neutron capture therapy.
Wyzlic IM and Soloway AH.
Tetrahedron Letters, 33(49), 7489-7490 (1992)
Tetrahedron Letters, 4625-4625 (1978)
J J Knittel et al.
Peptide research, 3(4), 176-181 (1990-07-01)
A convenient approach to the synthesis of ring-substituted phenylalanine amides is described. Phase transfer alkylation of N-(diphenylmethylene)amino acetonitrile or N-(diphenylmethylene)glycine ethyl ester provides alpha-substituted imines 2 and 6. After acid hydrolysis and esterification, resolution with alpha-chymotrypsin provided 3'-substituted phenylalanine analogs
Tetrahedron Letters, 48, 945-945 (2007)
Synthesis, 918-918 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service