Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

252409

Sigma-Aldrich

2,3,7,8,12,13,17,18-Octaethyl-21H,23H-porphine

97%

Synonym(s):

Octaethylporphine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H46N4
CAS Number:
Molecular Weight:
534.78
Beilstein:
379798
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

powder

λmax

401 nm

SMILES string

CCc1c(CC)c2cc3[nH]c(cc4nc(cc5[nH]c(cc1n2)c(CC)c5CC)c(CC)c4CC)c(CC)c3CC

InChI

1S/C36H46N4/c1-9-21-22(10-2)30-18-32-25(13-5)26(14-6)34(39-32)20-36-28(16-8)27(15-7)35(40-36)19-33-24(12-4)23(11-3)31(38-33)17-29(21)37-30/h17-20,37,40H,9-16H2,1-8H3/b29-17-,30-18-,31-17-,32-18-,33-19-,34-20-,35-19-,36-20-

InChI key

HCIIFBHDBOCSAF-MUZKIALCSA-N

Gene Information

human ... TERT(7015)

General description

Metalated and metal free 2,3,7,8,12,13,17,18-octaethyl- 21H,23H-porphine (OEP) can form Langmuir Blodgett (LB) thin films. OEP has gas sensing properties.1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service