Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

251879

Sigma-Aldrich

2,6-Dichlorophenyl isocyanate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3NCO
CAS Number:
Molecular Weight:
188.01
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

bp

101 °C/5 mmHg (lit.)

mp

42-44 °C (lit.)

SMILES string

Clc1cccc(Cl)c1N=C=O

InChI

1S/C7H3Cl2NO/c8-5-2-1-3-6(9)7(5)10-4-11/h1-3H

InChI key

HMVKMAMIRAVXAN-UHFFFAOYSA-N

Application

2,6-Dichlorophenyl isocyanate has been used in the preparation of pyridine-urea analog.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luping Liu et al.
Bioorganic & medicinal chemistry letters, 13(22), 3979-3982 (2003-11-01)
Development for a class of potent 3,4-dihydropyrido(3,2-d)pyrimidone inhibitors of p38a MAP kinase is described. Modification of N-1 aryl and C-6 arylsulfide in 3,4-dihydropyrido(3,2-d)pyrimidone analogues for the interaction with the hydrophobic pockets in p38 active site is also discussed.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service