Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

251356

Sigma-Aldrich

Methylenecyclopentane

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C5H8(=CH2)
CAS Number:
Molecular Weight:
82.14
Beilstein:
1847501
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.435 (lit.)

bp

75-76 °C (lit.)

density

0.781 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C=C1CCCC1

InChI

1S/C6H10/c1-6-4-2-3-5-6/h1-5H2

InChI key

NFJPEKRRHIYYES-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hydration rate of methylenecyclopentane has been measured spectrophotometrically in aqueous perchloric acid. Trost′s palladium-mediated methylenecyclopentane annelation to the nitroalkene yields methylenecyclopentane derivative.

Application

Methylenecyclopentane has been used in total synthesis of (±)-cephalotaxin.

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-0.4 °F - closed cup

Flash Point(C)

-18 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The total synthesis of cephalotaxine.
J Auerbach et al.
Journal of the American Chemical Society, 94(20), 7172-7173 (1972-10-04)
COMPARISON OF THE TRANSITION-STATE ENERGIES OF ACID-CATALYZED HYDRATIONS OF SOME 1-METHYLCYCLOALKENES AND METHYLENECYCLOALKANES.
Lajunen M and Himottu M.
Acta Chemica Scandinavica, 41(5), 252-258 (1987)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service